Learn More
Medchemexpress LLC Sodium carboxymethylcellulose | 9004-32-4 | MFCD81472 | 99.5% | 90,000 (average) | N/A | 5g
SDP
Supplier:  Medchemexpress LLC HYY0703D5G
Encompass_Preferred
Sodium carboxymethyl cellulose (average Mw 90000 DS 0 7 Viscosity 50-200 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent[1][2][3]
Catalog No. 50-002-69828