Learn More
Medchemexpress LLC Sodium caseinates | 9005-46-3 | 25 G
Supplier: Medchemexpress LLC 25GHY139410
Sodium caseinates are the salt of casein and the main milk protein. They are able to induce the proliferation and activation of granulocytes, increase the serum concentration of two key cytokines (GM-CSF and G-CSF), and induce mouse granulopoiesis.
- Salt of casein
- Main milk protein
- Induces proliferation and activation of granulocytes
- Increases serum concentration of GM-CSF and G-CSF
- Induces mouse granulopoiesis