Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium chlorate ACS reagen500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 403016500G
Encompass
Sodium chlorate is a hygroscopic achiral ionic compound which on crystallization forms two enantiomeric chiral crystals. It is reported to be a probable byproduct of the drinking water disinfectant chlorine dioxide (ClO2). d-sodium chlorate may be utilized as chiral initiators in the synthesis of (S)-5-pyrimidyl alkanols. Commercial production of sodium chlorate is by anodic oxidation of NaCl. It finds application as herbicide oxidizing agent and as a starting material in the synthesis of sodium perchlorate.
Catalog No. 50-289-7781