Learn More
Alkali Scientific Sodium Chloride, 1 Kg
SDP
Supplier:  Alkali Scientific ODI001CP
Encompass_Preferred
Sodium Chloride (NaCl), laboratory grade, is a fundamental chemical reagent used in buffer preparation and various laboratory applications. Available in a 1kg pack, it is commonly used in the preparation of physiological saline solutions, which are crucial for biological research, cell culture, and molecular biology. Sodium chloride is used to maintain osmotic balance and stabilize proteins, cells, and other biomolecules during experiments. It is also a key ingredient in preparing isotonic solutions that mimic the body's natural conditions for various assays. In addition, sodium chloride is widely used in the pharmaceutical and food industries as a preservative.
Catalog No. 50-006-60911