Learn More
ALADDIN SCIENTIFIC CORPORATION Sodium chloride, 100g
Supplier:  ALADDIN SCIENTIFIC CORPORATION C111549100G
Encompass_Preferred
Sodium Chloride (NaCl) is widely used in biochemistry and molecular biology applications It is found in nature in all body tissue and is considered an essential nutrient Used in a wide variety of biochemical applications including creating density gradients regulating biological functions and as a diluent to increase ionic strength in buffers or culture media NaCl is a component of phosphate buffered saline (PBS)and SSC buffer It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations The use of sodium chloride for isolation of DNA from mammalian cells has been described Recent studies provide evidence that a high amount of NaCl reduces production of nitric oxide Usually used in biochemistry and molecular biology applications a component of PBS and SSC buffers
Catalog No. 50-000-82530