Learn More
Hach Company Sodium Chloride Standard Solution, 100 µS/cm, 500 mL
Supplier:  Hach Company 2971849
Encompass
For accuracy checks in conductivity measurements. 49 mg/L as NaCl. 100 µS/cm @ 25°C. 500 mL.
Catalog No. 50-199-6976