Learn More
U.S. Pharmacopeia Sodium Chloride, 7647-14-5, MFCD00003477, 1g
Supplier:  U.S. Pharmacopeia 1613804
Encompass
Molecular formula NaCl, Molecular weight 58.44, Melting Point 1473.8 - 1479.2 °F (801 - 804 °C), Boiling Point 2575.4 °F (1413 °C)
Catalog No. NC0508307