Learn More
Research Products International Corp SODIUM CHLORIDE, 2.5KG
SDP
Supplier:  Research Products International Corp S230202500.0
Encompass_Preferred
Sodium chloride (NaCl) is a highly soluble, white crystalline salt (58.44 g/mol) used in life science labs to maintain osmotic balance and ionic strength. It is essential in buffers, cell culture media, nucleic acid extraction, protein purification, and electrophoresis, where consistent ionic conditions support biochemical and molecular processes.
Catalog No. 50-489-004