Learn More
Alkali Scientific Sodium Chloride, 25 Kg
SDP
Supplier:  Alkali Scientific ODI005CP
Encompass
Sodium Chloride, laboratory grade, is a critical reagent used in the preparation of buffers, saline solutions, and other chemical formulations in scientific laboratories. This 25 kg bulk pack ensures a large supply of Sodium Chloride, ideal for high-volume applications in research and industry. Sodium Chloride is used extensively in biological, pharmaceutical, and chemical processes, particularly where osmotic balance and solution stability are critical. It is widely used for the preparation of isotonic solutions, laboratory reagents, and in medical applications. The 25 kg packaging offers convenience for high-demand facilities requiring large quantities of high-purity Sodium Chloride.
Catalog No. 50-006-61374