Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium chloride, 7647-14-5, MFCD00003477, 5 kg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S76535KG
Encompass
Linear Formula: NaCl, Molecular Weight: 58.44, BioXtra, ≥99.5% (AT), Synonym: Halite. Sodium chloride has been used to: Prepare PBS (Phosphate-buffered saline) solution. Prepare lysis buffer.
Catalog No. NC1188618