Learn More
Quality Biological Inc Sodium Chloride 5 M, 500 mL
SDP
Supplier:  Quality Biological Inc 351036101
Encompass_Preferred
Sodium Chloride, 5M is suitable for molecular biology, protein chemistry and biochemical applications. ¿ 5 M concentration ¿ Filtered through a 0.22 ¿m membrane ¿ DNase, RNase, and Protease tested ¿ Applications: Sodium Chloride, 5 M is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase, and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 41105514 ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7303