Learn More
Boston Bioproducts Inc Sodium Chloride (5 M) - 500ml
Supplier:  Boston Bioproducts Inc BM244500ML
Encompass
Sodium Chloride, also refered to as saline, is a solution consisting of dissolved sodium chloride salt in varying sodium (Na+) and Chloride (Cl-) concentrations. This solution creates isotonic environments that mimic saline concentrations within living organisms. Sodium Chloride is a stable solution that provides a physiologicly neutral pH to experiments, making it very useful when combined with a buffer solution, or for use in cell culture. This product is formulated at 5M concentration.
Catalog No. NC9385658