Learn More
Alkali Scientific Sodium Chloride 5M Solution, 250 Milliliters
Supplier: Alkali Scientific IDE002CP
Sodium Chloride 5M Solution is a highly concentrated sodium chloride solution that is commonly used in laboratory and industrial applications requiring high salt concentrations. The 250mL bottle is designed for precise laboratory work, providing a consistent and stable source of sodium chloride for various biochemical experiments. It is often used in molecular biology for preparing buffers, solutions for electrophoresis, and dilutions in applications like protein or nucleic acid analysis. This high-concentration solution is also utilized in medical and clinical diagnostics for assays that require precise ionic strength, as well as in manufacturing processes where controlled salt levels are essential.