Learn More
Alkali Scientific Sodium Chloride 5M Solution, 500 Milliliters
SDP
Supplier:  Alkali Scientific IDE001CP
Encompass_Preferred
Sodium Chloride 5M Solution (500 mL) is a laboratory-grade reagent used in various biochemical and molecular biology applications. The high concentration of sodium chloride in this solution makes it ideal for preparing buffers, maintaining osmotic balance, and stabilizing proteins and other biomolecules. The 5M concentration is particularly useful in DNA extraction, protein purification, and enzyme assays, where a high ionic strength is required. This 500 mL bottle is convenient for medium to large-scale applications in laboratory settings, providing the required concentration of sodium chloride for various types of chemical and biological testing.
Catalog No. 50-006-59680