Learn More
Apexbio Technology LLC Sodium chloride 7647-14-5 1g
SDP
Supplier:  Apexbio Technology LLC B72881000
Encompass_Preferred
Sodium chloride (7647-14-5) is an inorganic salt that readily dissociates into sodium and chloride ions supporting essential cellular processes by maintaining osmotic balance and influencing cellular signaling pathways In experimental applications sodium chloride functions as a buffering and isotonic agent regulating osmotic pressure and supporting stable conditions for cell growth and differentiation in vitro It is routinely incorporated into media formulations buffer solutions and drug screening protocols Based on these properties sodium chloride holds research potential in basic biomedical studies pharmacological research and cell biology investigations
Catalog No. 50-000-89000