Learn More
Strem, An Ascensus Company CAS# 7647-14-5. 1kg. Sodium chloride, 99+% (ACS). MFCD00003477
SDP
Supplier:  Strem, An Ascensus Company 9311281000G
Encompass
CAS# 7647-14-5. 1kg. Sodium chloride, 99+% (ACS). MFCD00003477. Molecular Weight: 58.44. Molecular Formula: NaCl. Color/form: white xtl. Strem# 93-1128. http://www.strem.com/catalog/v/93-1128/
Catalog No. 50-901-13851