Learn More
Alkali Scientific Sodium Chloride, ACS Grade, 10 Kg
SDP
Supplier:  Alkali Scientific ODI009CP
Encompass_Preferred
Sodium Chloride, ACS grade, is a high-purity reagent that is critical in buffer preparation, saline solution creation, and various chemical reactions in laboratory settings. This 10 kg pack ensures a steady supply of high-grade Sodium Chloride for labs that require consistent, reliable quality. It is used in biological, pharmaceutical, and chemical research for creating isotonic solutions that maintain osmotic balance and ensure cell viability. The 10 kg packaging is ideal for high-demand laboratories where large quantities of Sodium Chloride are needed for extensive buffer preparation and sample preparation in various research applications.
Catalog No. 50-006-61397