Learn More
Crescent Chemical Co Inc SODIUM CHLORIDE MOL BIO GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE39781.02
Encompass_Preferred
Sodium chloride molecular biology grade DNase/RNase not detected. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 39781.02 CAS 7647-14-5
Catalog No. 50-248-468