Learn More
Bioworld Sodium Chloride Peptone Broth, 500 g
Supplier:  Bioworld 306201151
Encompass_Preferred
For preparation of microbial suspension, dilution or washing. The low content of peptone along with phosphate buffer and sodium chloride maintains the viability of micro organisms without supporting growth.Composition (g/L):3.6g Potassium phosphate, dibasic7.2g Sodium Phosphate, monobasic 4.3g Sodium chloride & 1g Enzymatic digest of CaseinStorage: Store cool at room temperature.
Catalog No. 50-254-419