Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium chloride solution technical, ~26% (saturated in water at 20C, AT) | 7647-14-5 | MFCD00003477 | 1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 713921L
Encompass
Sodium chloride solution technical, ~26% (saturated in water at 20C, AT) | Purity: ~26% (saturated in water at 20C, AT) | Mol Wt: 58.44 | 7647-14-5 | MFCD00003477 | 1L
Catalog No. 50-188-3796