Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium cyanide ACS reagent25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2052222.5KG
Encompass
Sodium cyanide is colorless sodium salt and its crystals belong to the cubic crystal system. Its aqueous SOLUTIONons are strongly basic due to the presence of cyanide ion. It can be synthesized by reacting sodium hydroxide with hydrogen cyanide. Sodium cyanide is a key precursor for the synthesis of various organic intermediates. It participates in the RuCl3-mediated oxidative cyanation of tertiary amines.
Catalog No. 50-289-8752