Learn More
Medchemexpress LLC Sodium diatrizoate | 737-31-5 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0926A10MM/1ML
Encompass_Preferred
Sodium diatrizoate is an iodinated radiocontrast agent. It has potential for radiographic imaging of the airways and induces mitochondrial turnover and oxidative stress, activating apoptosis by dysregulating calcium.
  • Iodinated radiocontrast agent
  • Potential for radiographic imaging of the airways
  • Induces mitochondrial turnover and oxidative stress
  • Activates apoptosis by dysregulating calcium
Catalog No. 50-004-09083