Learn More
Medchemexpress LLC Sodium dihydrogen phosphate dihydrate | 13472-35-0 | MFCD00149209 | 99.2% | 156.01 g/mol | NaH2PO4·2H2O | 1kg
SDP
Supplier:  Medchemexpress LLC HYY0332C1KG
Encompass_Preferred
Sodium dihydrogen phosphate dihydrate 99 0% (Sodium phosphate dihydrate 99 0%) is used as a buffer capacity reagent in molecular biology biochemistry and chromatography Sodium dihydrogen phosphate dihydrate 99 0% is also used as a laxative and pH buffer when combined with other sodium phosphates and also used as fuel and detergent additives[1]
Catalog No. 50-002-69334