Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium dodecyl sulfate, 151-21-3, MFCD00036175, 250 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 75746250G
Encompass
Linear Formula: CH3(CH2)11OSO3Na, Molecular Weight: 288.38, ≥99.0% (GC), dust-free pellets, Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate.
Catalog No. NC1188378