Learn More
Research Products International Corp SDS, Micro-Pellets [Sodium dodecylsulfate][Lauryl sulfate], 1 Kilogram
SDP
Supplier:  Research Products International Corp L220401000.0
Encompass_Preferred
New micro-pellets allow convenient weighing and handling without the mess and odor of? powders. High purity SDS is additive free. Easy to dissolve in water for denaturing proteins prior to gel elecrophoresis.
Catalog No. 50-488-739