Learn More
Strem, An Ascensus Company CAS# 141-52-6. 50g. Sodium ethoxide, 96%. MFCD00012417
SDP
Supplier:  Strem, An Ascensus Company 93113950G
Encompass
CAS# 141-52-6. 50g. Sodium ethoxide, 96%. MFCD00012417. Molecular Weight: 68.05. Molecular Formula: NaOC2H5. Color/form: white to yellow pwdr. Strem# 93-1139. http://www.strem.com/catalog/v/93-1139/
Catalog No. NC9616183