Learn More
Strem, An Ascensus Company CAS# 10163-15-2. 250g. Sodium fluorophosphate, 94%. MFCD00014248
SDP
Supplier:  Strem, An Ascensus Company 931059250G
Encompass
CAS# 10163-15-2. 250g. Sodium fluorophosphate, 94%. MFCD00014248. Molecular Weight: 143.95. Molecular Formula: Na2PO3F. Color/form: white pwdr. Strem# 93-1059. http://www.strem.com/catalog/v/93-1059/
Catalog No. 50-901-13806