Learn More
Medchemexpress LLC Sodium Glucoheptonate | 31138-65-5 | 98.0% | 248.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY136201
Encompass_Preferred
Sodium Glucoheptonate is used as a kit for radio-labeling with Tc-99m. Tc-99m glucoheptonate is a kidney- and brain-imaging agent, and can be used in brain-tumor detection. It exhibits rapid blood clearance and a quite stable blood level.
  • Used as a kit for radio-labeling with Tc-99m
  • Can be used in brain-tumor detection
  • Exhibits rapid blood clearance
  • Has a stable blood level
Catalog No. 50-003-51138