Learn More
Medchemexpress LLC Sodium glycolate | 2836-32-0 | MFCD00065358 | 98.0% | 98.03 g/mol | C2H3NaO3 | 25 G

Supplier: Medchemexpress LLC 25GHYW088011B
Sodium glycolate is the sodium salt of glycolic (hydroxyacetic) acid supplied as a white to off-white solid with 98.0% purity. It is used as a metal chelating agent and as a reagent in biochemical assays and buffer formulations.
- Used as a metal chelating agent in biochemical applications.
- Purity: 98.0%.
- CAS number: 2836-32-0.
- Chemical formula: C2H3NaO3.
- Molecular weight: 98.03 g/mol.
- Available in 10 g, 25 g, 50 g, and larger package sizes.
- Documents available: SDS, COA, data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.