Learn More
Strem, An Ascensus Company CAS# 14434-22-1. 500g. Sodium hexacyanoferrate(II) decahydrate. MFCD00149274
SDP
Supplier:  Strem, An Ascensus Company 931140500G
Encompass
CAS# 14434-22-1. 500g. Sodium hexacyanoferrate(II) decahydrate. MFCD00149274. Molecular Weight: 303.88 (484.04). Molecular Formula: Na4Fe(CN)6∙10H2O. Color/form: pale yellow xtl. Strem# 93-1140. http://www.strem.com/catalog/v/93-1140/
Catalog No. 50-901-13860