Learn More
TARGETMOL CHEMICALS INC SODIUM HYDROGEN SULFIDE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3650350MG
Encompass_Preferred
Also available in and bulk. Please contact Fisher for quotes. Sodium Hydrogen Sulfide (hydrate) is like nitric oxide an important gaseous mediator that has significant effects on the immunological neurological cardiovascular and pulmonary systems of mammals. Sodium Hydrogen Sulfide (hydrate) is an H2S donor commonly used in cellular and whole animal experimental systems. For example it has been used to suggest that H2S promotes neutrophil migration reduces airway inflammation and protects neurites heart and intestine from chemical or ischemic-reperfusion damage. purity: 98%
Catalog No. 50-258-0181