Learn More
Strem, An Ascensus Company Sodium bisulfide anhydrous 1g
SDP
Supplier:  Strem, An Ascensus Company 9310611G
Encompass
CAS# 16721-80-5. 1g. Sodium hydrogen sulfide, anhydrous. MFCD00011124. Molecular Weight: 56.06. Molecular Formula: NaHS. Color/form: off-white pwdr. Strem# 93-1061. http://www.strem.com/catalog/v/93-1061/
Catalog No. 50-901-13808