Learn More
Medchemexpress LLC Sodium Iodide | 7681-82-5 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB177010MM/1ML
Encompass_Preferred
Sodium Iodide is a compound composed of sodium ions and iodine ions. It scavenges hydrogen peroxide, maintains the level of thyroid hormone T3 through its iodine component, protects tissues from oxidative damage, and regulates thyroid homeostasis. It also relieves acute myocardial infarction.
  • Scavenges hydrogen peroxide.
  • Maintains the level of thyroid hormone T3 through its iodine component.
  • Protects tissues from oxidative damage and regulates thyroid homeostasis.
  • Relieves acute myocardial infarction.
  • In vivo, Sodium Iodide (1 mg/kg; i.v.) significantly reduces infarct size, decreases cardiac troponin levels, and maintains heart function in rat and pig models of acute myocardial infarction.
Catalog No. 50-004-12658