Learn More
Medchemexpress LLC Sodium Iodide | 7681-82-5 | 99.6% | 100 MG

Supplier: Medchemexpress LLC HYB1770100MG
Sodium Iodide is a compound composed of sodium ions and iodine ions. It scavenges hydrogen peroxide, maintains the level of thyroid hormone T3 through its iodine component, protects tissues from oxidative damage, regulates thyroid homeostasis, and relieves acute myocardial infarction.
- Molecular weight: 149.89
- Formula: INa
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: RT, protect from light, stored under nitrogen. In solvent: -80°C for 1 year, -20°C for 6 months
- Solubility in H2O: 200 mg/mL (1334.31 mM; need ultrasonic)
- Solubility in DMSO: 100 mg/mL (667.16 mM; ultrasonic and warming and heat to 60°C; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.