Learn More
Medchemexpress LLC Sodium Iodide | 7681-82-5 | 99.6% | 50 MG

Supplier: Medchemexpress LLC HYB177050MG
Sodium Iodide is a compound composed of sodium ions and iodine ions. It scavenges hydrogen peroxide, maintains thyroid hormone T3 levels through its iodine component, protects tissues from oxidative damage, regulates thyroid homeostasis, and relieves acute myocardial infarction. It is for research use only.
- Appearance: Solid
- Color: White to off-white
- Molecular Weight: 149.89
- Formula: INa
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: RT, protect from light, stored under nitrogen. In solvent: -80°C 1 year, -20°C 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.