Learn More
Medchemexpress LLC Sodium ionophore III | 81686-22-8 | MFCD00077941 | 99.9% | 552.8 g/mol | C34H52N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10110950MG
Encompass_Preferred
Sodium ionophore III is a sodium-selective ionophore used to determine sodium activity in biological samples such as blood, plasma, and serum. It is supplied as a high-purity research reagent suitable for assay development and analytical applications.
  • High purity (≈99.9%) supporting sensitive analytical measurements.
  • Selective transport of Na+ for reliable sodium activity assays.
  • Suitable for use in blood, plasma, and serum sample testing.
  • Molecular formula C34H52N2O4 and molecular weight 552.8 g/mol.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-13491