Learn More
Gelest Inc SODIUM ISOPROPOXIDE 100G
Supplier:  Gelest Inc AKS751100G
Encompass
This item has a minimum qty of 13 per supplier requirements.
C3H7NAO, MW: 82.08, SPECIFIC GRAVITY: 0.90, Sodium isopropoxide, 20% in tetrahydrofuran, CAS: 683-60-3
Catalog No. NC1767559