Learn More
Medchemexpress LLC Sodium lauroyl glutamate | 29923-31-7 | 95.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130313100MG
Encompass_Preferred
Sodium lauroyl glutamate is an anionic amino acid surfactant. It has the potential for irritant contact dermatitis and possible anti-irritating potential in a surfactant mixture on human skin. It is primarily used in cleaning products and has been shown to exhibit low irritation to nude mouse skin at 1% w/w for 1 hour. This product is for research use only and not sold to patients.
  • Anionic amino acid surfactant.
  • Low irritation to nude mouse skin at 1% w/w for 1 hour.
  • Primarily used in cleaning products.
  • Mild surfactant with a pH value of 5.41.
  • For research use only.
Catalog No. 50-003-38652