Learn More
Medchemexpress LLC Sodium lauroyl glutamate | 29923-31-7 | 95.81% | 10 G
SDP
Supplier:  Medchemexpress LLC HY13031310G
Encompass_Preferred
Sodium lauroyl glutamate is an anionic amino acid surfactant. It has been noted for its potential to cause irritant contact dermatitis, but also for its possible anti-irritating potential when part of a surfactant mixture on human skin. As a mild surfactant, it is primarily utilized in cleaning products. It has a pH value of 5.41 and demonstrates low irritation to the nude mouse skin.
  • Anionic amino acid surfactant
  • Used in cleaning products
  • Exhibits low irritation to nude mouse skin in vitro
  • For research use only; not sold to patients
Catalog No. 50-003-38423