Learn More
Medchemexpress LLC Sodium lauroyl glutamate | 29923-31-7 | 95.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY130313500MG
Encompass_Preferred
Sodium lauroyl glutamate is an anionic amino acid surfactant primarily used in cleaning products. While it has the potential for irritant contact dermatitis, it also exhibits possible anti-irritating properties in surfactant mixtures on human skin. A 1% w/w solution demonstrates low irritation and has a pH value of 5.41.
  • Anionic amino acid surfactant
  • Mild surfactant
  • Used mainly in cleaning products
  • Exhibits possible anti-irritating potential in surfactant mixtures
  • Low irritation to nude mouse skin at 1% w/w solution
  • 1% w/w solution has a pH value of 5.41
  • Appears as a white to off-white solid
Catalog No. 50-003-38429