Learn More
Medchemexpress LLC Sodium lauroylsarcosinate | 137-16-6 | MFCD00042728 | 99.7% | 293.38 g/mol | C15H28NNaO3 | 25G
SDP
Supplier:  Medchemexpress LLC HY12592025G
Encompass_Preferred
Sodium lauroylsarcosinate is the sodium salt of N-lauroylsarcosine, an anionic surfactant supplied as a white to off-white solid with high purity. It is used as a detergent and biochemical reagent for membrane solubilization, as a milder alternative to SDS in protein work, and in formulation and personal care research. Typical solubility is ≈50 mg/mL in water (requires ultrasonic) and ≈5 mg/mL in DMSO (requires warming/heating to 60 °C).
  • High purity solid suitable for biochemical assays.
  • Anionic surfactant used for membrane solubilization.
  • Milder alternative to SDS for protein work.
  • Soluble in water (~50 mg/mL) with ultrasonic treatment.
  • Soluble in DMSO (~5 mg/mL) with warming to 60 °C.
  • Useful in formulation and personal care research.
Catalog No. 50-002-37830