Learn More
Strem, An Ascensus Company CAS# 6834-92-0. 500g. Sodium metasilicate, anhydrous, min. 95%. MFCD00003492
SDP
Supplier:  Strem, An Ascensus Company 931196500G
Encompass
CAS# 6834-92-0. 500g. Sodium metasilicate, anhydrous, min. 95%. MFCD00003492. Molecular Weight: 122.06. Molecular Formula: Na2SiO3. Color/form: white pwdr. Strem# 93-1196. http://www.strem.com/catalog/v/93-1196/
Catalog No. 50-901-13918