Learn More
Medchemexpress LLC Sodium metatungstate | 12141-67-2 | MFCD00061466 | 93.4% | 2986.01 g/mol | H2Na6O40W12 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY103259500MG
Encompass_Preferred
Sodium metatungstate is an inorganic polyoxometalate salt used in biochemical research as a reagent and NTPDase inhibitor. It is supplied as a white to off-white solid with defined assay and solubility properties for laboratory applications.
  • White to off-white solid with an assay of 93.4%.
  • Soluble in water and DMSO; aqueous solubility up to 50 mg/mL (may require ultrasonic).
  • Available in multiple package sizes, including 500 MG.
  • Store sealed at 4 °C; in solution store at -80 °C (6 months) or -20 °C (1 month).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-47573