Learn More
Medchemexpress LLC Sodium methanesulfonate | 2386-57-4 | MFCD00064389 | 98.0% | 118.09 g/mol | CH3NaO3S | 500 G

Supplier: Medchemexpress LLC HYW034344500G
Sodium methanesulfonate is a white crystalline sodium salt used as a reagent and buffer component in biochemical and life-science research. It is supplied at laboratory grade and intended for research use; typical applications include buffer formulation, reagent preparation, and use in analytical workflows where a non-nucleophilic sulfonate counterion is required.
- High purity (98.0%) suitable for research applications.
- Molecular formula CH3NaO3S and molecular weight 118.09 g/mol.
- Soluble in DMSO and aqueous solvents for flexible assay use.
- Stable when stored sealed and protected from moisture.
- Available in multiple pack sizes, including 500 g, for bulk use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.