Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Sodium Methoxide, Anhydrous 25 GR
SDP

Supplier:  Electron Microscopy Sciences 21200

Encompass_Preferred

(Sodium Methylate)
CAS #124-41-4 CH₃Ona F.W. 54.03

Used in the removal of resin from sections.
pH: 5% solution @25ºC- pH 4.1-4.5

Catalog No. 50-287-43


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.