Learn More
Medchemexpress LLC Sodium methylarsonate | 2163-80-6 | MFCD00072437 | 95.0% | 161.95 g/mol | CH4AsNaO3 | 1mg
SDP
Supplier:  Medchemexpress LLC HYW7142251MG
Encompass_Preferred
Sodium methylarsonate (Monosodium methylarsonate) is an organoarsenic compound with environmental toxicity that can be used as a herbicide to control weeds Sodium methylarsonate interferes with plant metabolic processes particularly by hindering photosynthesis and protein synthesis ultimately leading to plant death Additionally sodium methylarsonate can induce the production of metallothioneins (MTs) in fish[1]
Catalog No. 50-002-70087