Learn More
Apexbio Technology LLC Sodium molybdate,5g CAS# 7631-95-0
SDP
Supplier:  Apexbio Technology LLC B78435000
Encompass_Preferred
Sodium molybdate is an acid phosphatase inhibitor. The osteoclastic acid phosphatase isoenzyme, secreted by osteoclasts, is a member of a widely-distributed class of iron-containing proteins with acid phosphatase activity. Elevated plasma isoenzyme levels are associated with increased bone turnover in metabolic disease. By inhibiting this class of acid phosphatases, sodium molybdate is capable of abolishing bone resorption. In addition, sodium molybdate has been shown to stabilize the nonactivated glucocorticoid-receptor complex.Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0516