Learn More
Strem, An Ascensus Company CAS# 10102-40-6. 500g. Sodium molybdate dihydrate, 99.5% (ACS). MFCD00149170
SDP
Supplier:  Strem, An Ascensus Company 424234500G
Encompass
CAS# 10102-40-6. 500g. Sodium molybdate dihydrate, 99.5% (ACS). MFCD00149170. Molecular Weight: 205.92 (241.95). Molecular Formula: Na2MoO4∙2H2O. Color/form: white pwdr. Strem# 42-4234. http://www.strem.com/catalog/v/42-4234/
Catalog No. 50-901-11721