Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium molybdate dihydrate100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 331058100G
Encompass
Sodium molybdate dihydrate (Na2MoO4.2H2O) is basic in nature. Its crystals belong to the orthorhombic crystal system and space group Pbca.
Catalog No. 50-289-8529