Learn More
Apexbio Technology LLC Sodium Nitroprusside 14402-89-2 50mg
SDP
Supplier:  Apexbio Technology LLC B202650
Encompass_Preferred
Sodium nitroprusside is an inorganic nitric oxide (NO) donor molecule used to modulate vascular tone and platelet responses It exerts its biological activity primarily through the release of NO an endogenous signaling molecule which induces vasodilation and inhibits platelet aggregation by interfering with calcium influx in vascular smooth muscle cells and reducing ADP and ATP release from activated platelets Based on these pharmacological properties sodium nitroprusside holds research potential in cardiovascular function studies vascular tone modulation and investigation of platelet responses
Catalog No. 50-000-93291